missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXC-ELR Antibody (Lilly patent anti-Pan-ELR+), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28036AF350
This item is not returnable.
View return policy
Description
CXC-ELR Monoclonal antibody specifically detects CXC-ELR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXC-ELR | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Lilly patent anti-Pan-ELR+ | |
| 50mM Sodium Borate | |
| CXC-ELR | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction