missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL1/2/3/GRO Antibody (29702), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2761S
This item is not returnable.
View return policy
Description
CXCL1/2/3/GRO Monoclonal antibody specifically detects CXCL1/2/3/GRO in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| CXCL1/2/3/GRO | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2919 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 29702 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
| E. coli-derived recombinant human CXCL3, Ala35-Asn107, Accession # P19876 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction