missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL1/GRO alpha/KC/CINC-1 Antibody (1174A), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB4532K
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal antibody specifically detects CXCL1/GRO alpha/KC/CINC-1 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining) | |
| 1174A | |
| Intracellular Staining by Flow Cytometry | |
| FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
| E. coli-derived recombinant mouse CXCL1/GRO alpha/KC/CINC-1, Asn29-Lys96, Accession # P12850 | |
| 0.1 mL | |
| Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Membrane Trafficking and Chaperones, Signal Transduction | |
| 2919 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction