missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL10/IP-10/CRG-2 Antibody (6D4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59749AF750
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Monoclonal antibody specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| CXCL10/IP-10/CRG-2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| C7, chemokine (C-X-C motif) ligand 10, crg-2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
| Recombinant human CXCL10/INP10 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 3627 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 6D4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction