missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL11/I-TAC Antibody (10C6), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22533UV
This item is not returnable.
View return policy
Description
CXCL11/I-TAC Monoclonal specifically detects CXCL11/I-TAC in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CXCL11/I-TAC | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CXCL11 | |
| Full length recombinant protein expressed in E-Coli | |
| 0.2 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| 6373 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 10C6 | |
| Western Blot, ELISA | |
| Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction