missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL11/I-TAC Antibody (MM0210-9N26), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-12219CL1
This item is not returnable.
View return policy
Description
CXCL11/I-TAC Monoclonal antibody specifically detects CXCL11/I-TAC in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
Specifications
| CXCL11/I-TAC | |
| Monoclonal | |
| CoraFluor 1 | |
| Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
| Human recombinant CXCL11/I-TAC | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| 6373 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Immunohistochemistry, Western Blot, Neutralization | |
| MM0210-9N26 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction