missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 Antibody (MM0211-9N26), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12221AF594
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Rat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CXCL12/SDF-1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CXCL12 | |
| Human recombinant CXCL12/SDF-1 | |
| 0.1 mL | |
| Primary | |
| Detects human CXCL12 (both 1-alpha and 1 beta) | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0211-9N26 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
| Mouse | |
| Protein G purified | |
| Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers | |
| 6387.0 | |
| Human, Rat, Sheep | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction