missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 Antibody (MM0211-9N26), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12221AF700
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Rat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CXCL12/SDF-1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Q6ICW0 | |
| CXCL12 | |
| Human recombinant CXCL12/SDF-1 | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 6387 | |
| Human, Rat, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0211-9N26 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects human CXCL12 (both 1-alpha and 1 beta) | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction