missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL12/SDF-1 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05873-50ul
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 beta Polyclonal antibody specifically detects CXCL12/SDF-1 beta in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| CXCL12/SDF-1 beta | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| hSDF-1beta, SDF 1beta, SDF1B | |
| Produced in rabbits immunized with purified, recombinant Mouse CXCL12/SDF-1 beta (Uniprot#: P40224-1; Lys22-Lys89) | |
| 50 μg | |
| Biologically Active Proteins | |
| 6387 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction