Learn More
CXCL14/BRAK Antibody (MM0213-12B24), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-12224AF532
Product Code. 18015488
  • ONLINE EXCLUSIVE PRICE
    € 615.00
    € 581.70 / 0.10mL
    Save € 33.30
    5% Off
Description
CXCL14/BRAK Monoclonal specifically detects CXCL14/BRAK in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL14/BRAK
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
CXCL14
Human recombinant CXCL14/BRAK
0.1 mL
Primary
Human
IgG2
Western Blot, Immunohistochemistry
MM0213-12B24
Western Blot, Immunohistochemistry
BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14
Mouse
Protein G purified
Biologically Active Proteins
9547.0
Store at 4°C in the dark.