missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL14/BRAK Antibody (MM0213-12B24), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12224DL594
This item is not returnable.
View return policy
Description
CXCL14/BRAK Monoclonal antibody specifically detects CXCL14/BRAK in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CXCL14/BRAK | |
| Monoclonal | |
| DyLight 594 | |
| BMAC, BRAKKEC, chemokine (C-X-C motif) ligand 14, Chemokine BRAK, member 14 (BRAK), MIP-2 gamma, MIP2G, MIP-2G, NJACKec, SCYB14MGC10687, Small-inducible cytokine B14 | |
| Human recombinant CXCL14/BRAK | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 9547 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry | |
| MM0213-12B24 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction