missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Antibody (256213) [HRP], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB976H
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF
Specifications
| CXCL16 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Neutralization, CyTOF | |
| 256213 | |
| Western Blot, Flow Cytometry, Neutralization, CyTOF-ready | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| E. coli-derived recombinant human CXCL16, Asn30-Pro118, Accession # Q9H2A7.4 | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| 58191 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction