missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Antibody (RM0208-15Q2), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12226V
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal specifically detects CXCL16 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CXCL16 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CXCL16 | |
| Mouse recombinant CXCL16 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0208-15Q2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| Rat | |
| Protein A or G purified | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
| 58191 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction