Learn More
CXCL16 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29589-1-AP-150UL
 View more versions of this product
Product Code. 18049415
  • € 410.00 / 150µL
Description
The CXCL16 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL16. This antibody recognizes human antigen. The CXCL16 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CXCL16
800 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
C X C motif chemokine 16, CXCL16, CXCLG16, SCYB16, Small inducible cytokine B16, SR PSOX, SRPSOX, Transmembrane chemokine CXCL16
150 μL
58191
Store at -20°C. Stable for one year after shipment.