Learn More
CXCL16 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29589-1-AP-150UL
This item is not returnable.
View return policy
Description
The CXCL16 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CXCL16. This antibody recognizes human antigen. The CXCL16 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CXCL16 | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| C X C motif chemokine 16, CXCL16, CXCLG16, SCYB16, Small inducible cytokine B16, SR PSOX, SRPSOX, Transmembrane chemokine CXCL16 | |
| 150 μL | |
| 58191 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |