missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL4/PF4 Antibody (170138), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7952H
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal antibody specifically detects CXCL4/PF4 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 170138 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| E. coli-derived recombinant human CXCL4/PF4, Glu32-Ser101, Accession # P02776.2 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cardiovascular Biology, Signal Transduction | |
| 5196 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction