missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL4/PF4 Antibody (U.Penn. patent anti-PF4), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27973AF350
This item is not returnable.
View return policy
Description
CXCL4/PF4 Monoclonal antibody specifically detects CXCL4/PF4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCL4/PF4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| CXCL4/ PF4 | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer, Cardiovascular Biology, Signal Transduction | |
| 5196 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| U.Penn. patent anti-PF4 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction