missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL5/ENA-78 Antibody (32B3), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22026FR
This item is not returnable.
View return policy
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| CXCL5 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CXCL5 | |
| Mouse recombinant CXCL5/ENA-78 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| 32B3 | |
| Western Blot, ELISA | |
| chemokine (C-X-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), epithelial-derived neutrophil activating protein 78, epithelial-derived neutrophil-activating peptide 78, Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78, small inducible cytokine B5, small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derivedneutrophil-activating peptide 78), Small-inducible cytokine B5 | |
| Rat | |
| Protein G purified | |
| Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
| 6374 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction