Learn More
CXCL5/ENA-78 Antibody (33170), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB654P
Product Code. 30031061
  • € 663.00 / 0.10mL
Description
CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Human samples. It is validated for ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL5/ENA-78
Monoclonal
PE
PBS
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry
33170
ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78
E. coli-derived recombinant human CXCL5/ENA-78, Ala37-Asn114, Accession # P42830
0.1 mL
Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
6374
Store at 4C in the dark.