Learn More
Novus Biologicals™ CXCL5/ENA-78 Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
missing translation for 'unitSize'
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CXCL5/ENA-78.
Spécification
Spécification
| Assay Range | 3.91 pg/mL |
| Assay Sensitivity | 3.91-250 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CXCL5/ENA-78. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 |
| Gene ID (Entrez) | 6374 |
| Afficher plus |