missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL6/GCP-2 Antibody (MM0217-10L25), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12232FR
This item is not returnable.
View return policy
Description
CXCL6/GCP-2 Monoclonal specifically detects CXCL6/GCP-2 in Human samples. It is validated for Western Blot.
Specifications
| CXCL6/GCP-2 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CXCL6 | |
| Human recombinant CXCL6/GCP-2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0217-10L25 | |
| Western Blot | |
| chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2), Chemokine alpha 3, CKA-3Granulocyte chemotactic protein 2, GCP2member 6 (granulocytechemotactic protein 2), GCP-2Small-inducible cytokine B6, Small inducible cytokine subfamily B (Cys-X-Cys), member b | |
| Mouse | |
| Protein G purified | |
| Cell Cycle and Replication, Chemokines and Cytokines | |
| 6372 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction