missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL-8 Antibody (6217), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB208C
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Simple Western, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 6217 | |
| Western Blot, Simple Western, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
| E. coli-derived recombinant human IL-8/CXCL8, Ser28-Ser99, Accession # P10145 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 3576 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction