missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL-8 Antibody (HuMax-IL8), mFluor Violet 450 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28022MFV450
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
| CXCL8/ IL-8 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 3576 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| HuMax-IL8 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction