Learn More
Novus Biologicals™ CXCL9/MIG Antibody (Novimmune patent anti-CXCL9) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28413-100ug
 View more versions of this product
Product Code. 30045612
  • € 338.00 / 100µg
Description
CXCL9/MIG Monoclonal antibody specifically detects CXCL9/MIG in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL9/MIG
Monoclonal
Unconjugated
Q07325
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Novimmune patent anti-CXCL9
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
CXCL9
100 μg
Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
4283