missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR1/IL-8RA Antibody (MM0221-7D22), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12237APCCY7
This item is not returnable.
View return policy
Description
CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human samples. It is validated for Flow Cytometry.
Specifications
| CXCR1/IL-8RA | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CXCR1 | |
| Human recombinant CXCR1/IL-8RA | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Flow Cytometry | |
| MM0221-7D22 | |
| Flow Cytometry | |
| CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A | |
| Mouse | |
| Protein G purified | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 3577.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction