Learn More
Invitrogen™ CXCR1 Monoclonal Antibody (501)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1206
This item is not returnable.
View return policy
Description
MA1-206 detects CXCR1/IL8RA from human samples. MA1-206 has been successfully tested in western blot, immunofluorescence and flow cytometry applications. In western blot, MA1-206 detects CXCR1/IL8RA (human) at approximate molecular weight of 30 kDa.
CXCR1 is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. Knockout studies in mice suggested that this protein inhibits embryonic oligodendrocyte precursor migration in developing spinal cord. This gene, IL8RB, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36.
Specifications
| CXCR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4 | |
| P25024 | |
| CXCR1 | |
| Synthetic peptide corresponding to the N-terminal residues 5-19 of human CXCR1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 501 | |
| Unconjugated | |
| CXCR1 | |
| C Cmotif chemokine; C X C motif chemokine; C-C; CC motif chemokine; C-C-CKR-1; CCmotif chemokine; CD128; CD181; CDw128a; chemokine (C-X-C motif) receptor 1; CKR-1; CMKAR1; CXC; CXC chemokine receptor 1; C-X-C chemokine receptor type 1; CXC motif chemokine; C-X-C motif chemokine receptor 1; CXCR1; CXC-R1; CXCR-1; High affinity interleukin-8 receptor A; IL-8 receptor; IL-8 Receptor alpha; IL-8 Receptor IL-RA; IL-8 receptor type 1; IL-8R A; IL8R1; Il8ra; IL8RBA; IL-RA; interleukin 8 receptor; interleukin 8 receptor alpha; interleukin 8 receptor, alpha; interleukin-8 receptor type 1; interleukin-8 receptor type A | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3577 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |