Learn More
CXCR2/IL-8RB Antibody (866614R), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8110RY
Product Code. 18597939
  • € 679.00 / 0.10mL
Description
CXCR2/IL-8RB Monoclonal antibody specifically detects CXCR2/IL-8RB in Rat samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCR2/IL-8RB
Monoclonal
DyLight 680
50mM Sodium Borate
Mouse
Protein A or G purified from cell culture supernatant
RUO
Primary
Rat
Purified
Flow Cytometry, CyTOF
866614R
Flow Cytometry, CyTOF-ready
CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
NS0 mouse myeloma cell line transfected with rat CXCR2/IL-8 RB, Accession # P35407
0.1 mL
Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
3579
Store at 4°C in the dark.
IgG2b