Learn More
CXCR3 Antibody (220803R) [DyLight 680], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB1685RY
Product Code. 18530605
  • € 706.00 / 0.10mL
Description
CXCR3 Monoclonal antibody specifically detects CXCR3 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCR3
Monoclonal
DyLight 680
50mM Sodium Borate
Rat
Protein A or G purified from cell culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
220803R
Flow Cytometry, CyTOF-ready
CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR
CHO Chinese hamster ovary cell line transfected with mouse CXCR3, Met1-Leu367, Accession # AAC40163
0.1 mL
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
2833
Store at 4°C in the dark.