missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR4 Antibody (30L4), Alexa Fluor™ 700, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22006AF700
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CXCR4 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 30L4 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CXCR4 | |
| Mouse recombinant CXCR4 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
| 7852 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction