Learn More
CXCR4 Mouse anti-Human, Clone: 12G5, Abnova™
Mouse monoclonal antibody raised against human CXCR4.
Brand: Abnova MAB12537.50ug
Description
This gene encodes a CXC chemokine receptor specific for stromal cell-derived factor-1. The protein has 7 transmembrane regions and is located on the cell surface. It acts with the CD4 protein to support HIV entry into cells and is also highly expressed in breast cancer cells. Mutations in this gene have been associated with WHIM (warts, hypogammaglobulinemia, infections, and myelokathexis) syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Specifications
| CXCR4 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry Immunofluorescence Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD184/D2S201E/FB22/HM89/HSY3RR/LAP3/LCR1/LESTR/NPY3R/NPYR/NPYRL/NPYY3R/WHIM | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 7852 | |
| Store at 4°C. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
| 12G5 | |
| Mouse monoclonal antibody raised against human CXCR4. | |
| In PBS, pH 7.2 (0.09% sodium azide). | |
| CXCR4 | |
| cP-MAC-infected Sup-T1 cells | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.