Learn More
CXCR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11073-2-AP-150UL
 View more versions of this product
Product Code. 16857903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CXCR4 may be involved in WHIM syndrome (WHIMS). Immunodeficiency disease characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection. Despite the peripheral neutropenia, bone marrow aspirates from affected individuals contain abundant mature myeloid cells, a condition termed myelokathexis. The disease is caused by mutations affecting the gene represented in this entry.
Specifications
Specifications
CXCR4
Polyclonal
Unconjugated
CXCR4
CD184, CXC R4, CXCR 4, CXCR4, CXCR4B, D2S201E, FB22, Fusin, HM89, HSY3RR, LAP3, LCR1, LESTR, SDF 1 receptor, WHIM
Rabbit
Antigen Affinity Chromatography
RUO
12767, 60628, 7852
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08565, P61073, P70658
Cxcr4
CXCR4 Fusion Protein Ag1528
150 μL
Primary
Human, Mouse, Rat