missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR5 Antibody (51505), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB190E
This item is not returnable.
View return policy
Description
CXCR5 Monoclonal antibody specifically detects CXCR5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-reported
Specifications
| CXCR5 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF | |
| 51505 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, CyTOF-reported | |
| BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| NS0 mouse myeloma cell line transfected with human CXCR5, Met1-Phe372, Accession # P32302 | |
| 0.1 mL | |
| Adaptive Immunity, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| 643 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction