missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1385
This item is not returnable.
View return policy
Description
CXCR5 Polyclonal antibody specifically detects CXCR5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CXCR5 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) | |
| BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| Synthetic 18 amino acid peptide from C-terminus of human CXCR5. | |
| 0.05 mL | |
| Adaptive Immunity, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| 643 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 5 μg/mL | |
| P32302 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction