missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (970106), Alexa Fluor 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB13382AF405
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| alpha-Synuclein | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SNCA | |
| This alpha-Synuclein Antibody (970106) was developed against E. coli-derived recombinant human alpha -Synuclein. | |
| 100 μL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 970106 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction