missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR6 Antibody (SR1646), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | CXCR6 |
|---|---|
| Clone | SR1646 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624664
|
Novus Biologicals
NBP3-22635-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642675
|
Novus Biologicals
NBP3-22635-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR6 Monoclonal antibody specifically detects CXCR6 in Human samples. It is validated for Western BlotSpecifications
| CXCR6 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| A synthesized peptide derived from human CXCR6 (Uniprot #: O00574) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1646 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 10663 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title