missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CXCR7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527077
This item is not returnable.
View return policy
Description
Recommended positive controls: PC-3, U87-MG, human CXCR7 transfected Mesenchymal stem cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
Specifications
| CXCR7 | |
| Polyclonal | |
| Unconjugated | |
| Ackr3 | |
| ACKR3; Atypical chemokine receptor 3; AW541270; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine; chemokine (C-X-C motif) receptor 7; chemokine orphan receptor 1; Cmkor1; CXC; C-X-C chemokine receptor type 7; CXC motif chemokine; CXCR7; CXC-R7; CXCR-7; G protein-coupled receptor; GPR159; G-protein coupled receptor 159; G-protein coupled receptor RDC1 homolog; RDC1; RDC-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12778, 57007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.14 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P25106, P56485 | |
| Ackr3 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human CXCR7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction