missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR7/RDC-1 Antibody (358426R), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4227RMFV610
This item is not returnable.
View return policy
Description
CXCR7/RDC-1 Monoclonal antibody specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| CXCR7/RDC-1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chemokines and Cytokines, GPCR | |
| 57007 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 358426R | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362 (Gly131Ser), Accession # AAA62370 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction