missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXorf56 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24021-1-AP-150UL
This item is not returnable.
View return policy
Beschreibung
While this gene is well-supported by transcript data, no functional information on its protein products is currently available. Three transcript variants encoding different isoforms have been found for this gene.Spezifikation
| CXorf56 | |
| Polyclonal | |
| Unconjugated | |
| CXorf56 | |
| CXorf56, UPF0428 protein CXorf56 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 63932 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H5V9 | |
| CXorf56 | |
| CXorf56 Fusion Protein Ag19856 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur