missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92186-0.02ml
This item is not returnable.
View return policy
Description
CXX1 Polyclonal antibody specifically detects CXX1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| CXX1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CAAX box 1, Cerebral protein 5, CXX1MAR8, family with sequence similarity 127, member A, Mammalian retrotransposon derived protein 8C, Mar8, MAR8CMGC117411, MART8, MART8C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human FAM127A (NP_001071639.1). MDGRVQLIKALLALPIRPATRRWRNPIPFPETFDGDTDRLPEFIVQTGSYMFVDENTFSSDALKVTFLITRLTGPALQWVIPYIKKESPLLNDYRGFLAEMKRVFGWEEDEDF | |
| 0.02 mL | |
| Cell Biology | |
| 8933 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction