Learn More
Macherey-Nagel™ Cyantesmo Qualitative Test Paper
Brand: Macherey-Nagel™ 90604
Additional Details : Weight : 0.22680kg
Specifications
Pale Green to Blue | |
For detection of Hydrocyanic acid, Cyanides | |
Qualitative Test Paper |
Paper | |
1 Each |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Cyantesmo
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : N,N,N',N'-tetramethyl-4,4'-diaminodiphenylmethane