missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYB561 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15883014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15883014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15883014 Supplier Invitrogen™ Supplier No. PA568029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

Secretory vesicle-specific electron transport protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB561
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYB561
Gene Accession No. P49447, Q60720
Gene Alias CD158F; CD158F1; Cyb561; CYB561A1; cytochrome b561; Cytochrome b-561; cytochrome b561 family, member A1; ferric-chelate reductase 2; FRRS2; KIR2DL5; Mcyt
Gene Symbols CYB561
Host Species Rabbit
Immunogen Synthesized peptide derived from human CYB561.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13056, 1534
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.