missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB561D2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB300-913
This item is not returnable.
View return policy
Description
CYB561D2 Polyclonal specifically detects CYB561D2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| CYB561D2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| cytochrome b-561 domain containing 2,101F6cytochrome b561 domain-containing protein 2, putative tumor suppressor 101F6, Putative tumor suppressor protein 101F6, TSP10 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Cow and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:16000 | |
| NP_008953. | |
| CYB561D2 | |
| Peptide with sequence C-VSNAYLYRKRIQP corresponding to C-Terminus according to NP_008953.1. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 11068 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction