missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYB5R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550071
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total CYB5R1 protein.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
Specifications
| CYB5R1 | |
| Polyclonal | |
| Unconjugated | |
| CYB5R1 | |
| 1500005G05Rik; b5R.1; B5R1; B5R2; C80155; Cyb5r1; cytochrome b5 reductase 1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; cytochrome b5 reductase 1; NADH-cytochrome b5 reductase 1; Nqo3a2; UNQ3049/PRO9865 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 304805, 51706, 72017 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5EB81, Q9DB73, Q9UHQ9 | |
| CYB5R1 | |
| Synthesized peptide derived from internal of human CYB5R1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction