missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97413MFV450
This item is not returnable.
View return policy
Description
CYB5R2 Polyclonal antibody specifically detects CYB5R2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CYB5R2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| B5R.2, cytochrome b5 reductase 2 | |
| Produced in rabbits immunized with E. coli-derived Human CYB5R2 fragment. | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 51700 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction