missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYB5R2 Antibody (OTI4B9), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71942PEATT594
This item is not returnable.
View return policy
Description
CYB5R2 Monoclonal specifically detects CYB5R2 in Human samples. It is validated for Flow Cytometry.
Specifications
| CYB5R2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| CYB5R2 | |
| Full length recombinant protein of human CYB5R2 (NP_057313) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| OTI4B9 | |
| Flow Cytometry | |
| B5R.2, cytochrome b5 reductase 2 | |
| Mouse | |
| Affinity Purified | |
| Cardiovascular Biology | |
| 51700.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction