Learn More
CYB5R2 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503251S
This item is not returnable.
View return policy
Description
CYB5R2 Monoclonal antibody specifically detects CYB5R2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CYB5R2 | |
| Monoclonal | |
| 1.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q03154 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 95 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI4B9 | |
| Unconjugated | |
| ACY1 | |
| b5R.2; b5R2; CYB5R2; cytochrome b5 reductase 2; cytochrome b5 reductase b5R.2; D630003K02Rik; NADH-cytochrome b5 reductase 2; RGD1308421 | |
| Full length human recombinant protein of human ACY1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |