missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYB5R4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15808663
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15808663 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15808663 Supplier Invitrogen™ Supplier No. PA570250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

NCB5OR is a flavohemoprotein that contains functional domains found in both cytochrome b5 (CYB5A; MIM 250790) and CYB5 reductase (see MIM 250800).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYB5R4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Cyb5r4
Gene Accession No. Q3TDX8
Gene Alias 2810034J18Rik; b5&b5R; b5/b5r; B5+B5R; b5b5r; C79736; cb5/cb5R; Cb5cb5r; Cyb5r4; cytochrome b5 reductase 4; cytochrome b-type NAD(P)H oxidoreductase; dJ676J13.1; flavohemoprotein b5/b5R; flavohemoprotein b5+b5R; hypothetical protein LOC553685; im:4727699; NADPH cytochrome B5 oxidoreductase; Ncb5or; N-terminal cytochrome b5 and cytochrome b5 oxidoreductase domain-containing protein; RP4-676J13.1; Unknown (protein for MGC:134229); zgc:112177
Gene Symbols Cyb5r4
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse CYB5R4 (aa 5-54).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266690
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.