missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYBA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575653
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Cytochrome b is comprised of a light chain (alpha) and a heavy chain (beta). This gene encodes the light, alpha subunit which has been proposed as a primary component of the microbicidal oxidase system of phagocytes. Mutations in this gene are associated with autosomal recessive chronic granulomatous disease (CGD), that is characterized by the failure of activated phagocytes to generate superoxide, which is important for the microbicidal activity of these cells.
Specifications
| CYBA | |
| Polyclonal | |
| Unconjugated | |
| CYBA | |
| b558; CYBA; cytochrome b light chain; cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; cytochrome b-245 alpha chain; cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; cytochrome b558 alpha-subunit; cytochrome b558 subunit alpha; cytochrome beta-558; flavocytochrome b-558 alpha polypeptide; neutrophil cytochrome b 22 kDa polypeptide; nmf333; p22 phagocyte B-cytochrome; p22 phox; p22phox; p22-PHOX; superoxide-generating NADPH oxidase light chain subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13057, 1535, 79129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13498, Q61462, Q62737 | |
| CYBA | |
| Synthetic peptide corresponding to residues in Human CYBA. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction