missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclandelate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Cyclandelate
Specifications
Specifications
| Chemical Name or Material | Cyclandelate |
| CAS | 456-59-7 |
| Analyte or Component Names | Cyclandelate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H24 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mandelic acid, 3,3,5-trimethylcyclohexyl ester (6CI,7CI,8CI), Cyclohexanol, 3,3,5-trimethyl-, mandelate (8CI), 3,3,5-Trimethylcyclohexyl mandelate, 3,5,5-Trimethylcyclohexanol, mandelic acid ester, 3,5,5-Trimethylcyclohexyl amygdalate, 3,5,5-Trimethylcyclohexyl mandelate, Arto-Espasmol, BS 572, Capilan, Ciclospasmol, Clandilon, Cyclandelate, Cyclergine, Cyclobral, Cyclolyt, Cyclomandol, Cyclospasmol, Dilatan, Natil, Novodil, Perebral, Saiclate, Sancyclan, Sepyron, Spasmione, Spasmocyclon, Benzeneacetic acid, alpha-hydroxy-, 3,3,5-trimethylcyclohexyl ester, 3,3,5-Trimethylcyclohexanol alpha-phenyl-alpha-hydroxyacetate |
| InChI Formula | InChI=1S/C17H24O3/c1-12-9-14(11-17(2,3)10-12)20-16(19)15(18)13-7-5-4-6-8-13/h4-8,12,14-15,18H,9-11H2,1-3H3 |
| SMILES | CC1CC(CC(C)(C)C1)OC(=O)C(O)c2ccccc2 |
| IUPAC Name | (3,3,5-trimethylcyclohexyl) 2-hydroxy-2-phenylacetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction