missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1 Antibody (E43.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2659
This item is not returnable.
View return policy
Description
Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Human samples. It is validated for Western Blot.
Specifications
| Cyclin A2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| CCNA2 | |
| Human Cyclin A1 | |
| 0.1 mg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 890 | |
| Human | |
| Purified |
| Western Blot | |
| E43.2 | |
| Unconjugated | |
| CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cyclin A (E43.2). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction