missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1 Antibody (E70.1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2660MFV610
This item is not returnable.
View return policy
Description
Cyclin A1 Monoclonal antibody specifically detects Cyclin A1 in Bovine samples. It is validated for Western Blot
Specifications
| Cyclin A1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| Western Blot | |
| E70.1 | |
| Western Blot | |
| cyclin A1, cyclin-A1 | |
| Bovine Cyclin A1 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 8900 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction